Order Entry
Canada
Orders LinkContactUsLinkComponent
GTE buffer | Avantor

GTE buffer

100 mL
Catalog # CAAAJ62597-AE
Fournisseur #:  J62597-AE
GTE buffer
Catalog # CAAAJ62597-AE
CAS Number:  
GTE buffer

100 mL

CAS Number:  
Price
$54.80
Log in for availability
Each (100ml)
La quantité minimale de commande est de 1
Product Details & Documents

Storage temperature: 2 to 8°C

  • Suitable for miniprep and DNA purification

Contains 25 mM Tris, 50 mM glucose, and 10 mM EDTA.

Documents

Specifications

Specifications

  • Size:
    100 mL

Specifications