Order Entry
Canada
Orders LinkContactUsLinkComponent
MOPS, 0.5 M Buffer Solutions, Thermo Scientific | Avantor

MOPS, 0.5 M Buffer Solutions, Thermo Scientific

MOPS buffer solution, 0.5 M, 250 mL, 5.5
Catalog # CAAAJ62477-AK
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ62477-AK
Supplier:  
CAS Number:  
MOPS, 0.5 M Buffer Solutions, Thermo Scientific

MOPS buffer solution, 0.5 M, 250 mL, 5.5

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
3-MORPHOLINOPROPANESULFONIC AC Catalog #TCM0707-250G

Le produit TCM0707-250G est un remplacement de CAAAJ62477-AK et a le même emballage et les mêmes spécifications, mais peut être d'une quantité différente.

Product Details & Documents

Liquid

CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183

Specifications

Specifications

  • Concentration:
    0.5 M
  • Description:
    MOPS buffer solution
  • Size:
    250 mL
  • pH:
    5.5
  • Cat. No.:
    CAAAJ62477-AK
  • Odour/scent:
    Odorless
  • Storage temperature:
    2°C to 8°C
  • MDL No.:
    MFCD00006183

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.