RNA sample loading buffer, no ethidium bromide, Thermo Scientific
2 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J61937-S0
RNA sample loading buffer, no ethidium bromide, Thermo Scientific
Catalog # CAAAJ61937-S0
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
MDL No.: MFCD00213041
Applications: Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide
Documents
Specifications
Specifications
- Size:2 mL
- Cat. No.:CAAAJ61937-S0
- MDL No.:MFCD00213041
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



