Order Entry
Canada
Orders LinkContactUsLinkComponent
 

D(-)-Lactic acid

250 mg
Catalog # CAAAJ61871-MD
Corrosive
 :  J61871-MD
D(-)-Lactic acid
Catalog # CAAAJ61871-MD
CAS Number:  10326-41-7
D(-)-Lactic acid

250 mg

CAS Number:  10326-41-7
Price
643,85 $
Log in for availability
Each (250mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Useful chiral synthon, building block for depsipeptides

Powder

Useful chiral synthon, building block for depsipeptides

Synonyms:  (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
  • CAS Number:10326-41-7
  • Formula:C₃H₆O₃
  • MDL Number:MFCD00068313
  • Molecular Weight:90.08
  • Merck Index:14,5335
  • Storage Temperature:Refrigerator
  • EINECS:233-713-2
Specifications

Specifications

  • Size:
    250 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.