Order Entry
Canada
Orders LinkContactUsLinkComponent
MOPS, 0.5 M Buffer Solutions, Thermo Scientific

MOPS, 0.5 M Buffer Solutions, Thermo Scientific

MOPS buffer solution, 0.5 M, 100 mL, 8.0
Catalog # CAAAJ61236-AE
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ61236-AE
Supplier:  
CAS Number:  
MOPS, 0.5 M Buffer Solutions, Thermo Scientific

MOPS buffer solution, 0.5 M, 100 mL, 8.0

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MOPS 25GM Catalog #TCM0707-025G

Product TCM0707-025G is a replacement for CAAAJ61236-AE and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

Liquid

CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183

Specifications

Specifications

  • Concentration:
    0.5 M
  • Description:
    MOPS buffer solution
  • Size:
    100 mL
  • pH:
    8.0
  • Cat. No.:
    CAAAJ61236-AE
  • Odour/scent:
    Odorless
  • Storage temperature:
    2°C to 8°C
  • MDL No.:
    MFCD00006183

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.