Order Entry
Canada
Orders LinkContactUsLinkComponent
CHAPS Lysis Buffer (2X), Thermo Scientific

CHAPS Lysis Buffer (2X), Thermo Scientific

50 mL
Catalog # CAAAJ60812-AD
CHAPS Lysis Buffer (2X), Thermo Scientific
Catalog # CAAAJ60812-AD
Supplier:  
CAS Number:  
CHAPS Lysis Buffer (2X), Thermo Scientific

50 mL

CAS Number:  
Discontinued
Product Details & Documents

Liquid

Physical Form: Liquid
Appearance: Clear liquid
MDL No.: MFCD00282866

Documents

Specifications

Specifications

  • Size:
    50 mL
  • Cat. No.:
    CAAAJ60812-AD
  • MDL No.:
    MFCD00282866

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.