Order Entry
Canada
Orders LinkContactUsLinkComponent
CHAPS Lysis Buffer, Thermo Scientific

CHAPS Lysis Buffer, Thermo Scientific

100 mL
Catalog # CAAAJ60580-AE
Supplier #:  J60580-AE
CHAPS Lysis Buffer, Thermo Scientific
Catalog # CAAAJ60580-AE
CAS Number:  
CHAPS Lysis Buffer, Thermo Scientific

100 mL

CAS Number:  
Price
$351.10
Log in for availability
Each (100ml)
Minimum order quantity is 1
Product Details & Documents

Liquid

Physical Form: Liquid
Appearance: Clear liquid
MDL No.: MFCD00282866
Applications: For cell lysis and protein extraction

Documents

Specifications

Specifications

  • Size:
    100 mL
  • Cat. No.:
    CAAAJ60580-AE
  • MDL No.:
    MFCD00282866

Specifications