TBE sample buffer (6X), Thermo Scientific
50 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J60357-AD
TBE sample buffer (6X), Thermo Scientific
Catalog # CAAAJ60357-AD
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Blue
Documents
Specifications
Specifications
- Size:50 mL



