TBE sample buffer (6X), Thermo Scientific
25 mL
Supplier: Thermo Scientific Chemicals
: J60357-AC
TBE sample buffer (6X), Thermo Scientific
Catalog # CAAAJ60357-AC
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Blue
Specifications
Specifications
- Size:25 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



