MOPS, 0.5 M Buffer Solutions, Thermo Scientific
MOPS buffer solution, 0.5 M, 100 mL, 7.0
Supplier: Thermo Scientific Chemicals
: J60328-AE
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ60328-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183
Specifications
Specifications
- Concentration:0.5 M
- Description:MOPS buffer solution
- Size:100 mL
- pH:7.0
- Cat. No.:CAAAJ60328-AE
- Odour/scent:Odorless
- Storage temperature:2°C to 8°C
- MDL No.:MFCD00006183



