Orders LinkContactUsLinkComponent
Aphidicolin
1mg
  CAAAJ60236-MCR
 :  J60236-MCR
Aphidicolin
  CAAAJ60236-MCR
 :  38966-21-1
Aphidicolin
1mg
 :  38966-21-1
$350.80
Each (1mg)
Cyrstalline powder
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
 :  (3R,4R,4aR,6aS,8R,9R,11aS,11bS)-4,9-Bis(hydroxymethyl)-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta[a]naphthalene-3,9-diol
  •  :38966-21-1
  •  :C₂₀H₃₄O₄
  •  :MFCD00083214
  •  :338.488 g/mol
  •  :14,727
  •  :224 °C
  •  :Refrigerator
  • Size:
    1mg