Aphidicolin
5mg
Supplier: Thermo Scientific Chemicals
Supplier #: J60236-LB0
Jump to:
Product Details & Documents
Cyrstalline powder
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
Synonyms: (3R,4R,4aR,6aS,8R,9R,11aS,11bS)-4,9-Bis(hydroxymethyl)-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta[a]naphthalene-3,9-diol
- CAS Number:38966-21-1
- Formula:C₂₀H₃₄O₄
- MDL Number:MFCD00083214
- Molecular Weight:338.488 g/mol
- Merck Index:14,727
- Melting Pt:224 °C
- Storage Temperature:Refrigerator
Documents
Specifications
Specifications
- Size:5mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



