Denaturing lysis buffer, Thermo Scientific
500 mL
Denaturing lysis buffer, Thermo Scientific
Catalog # CAAAJ60132-AP
CAS Number:
Jump to:
Discontinued
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Clear liquid
pH: 7.4
Documents
Specifications
Specifications
- Size:500 mL



