Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Schott Blue, GL45
Catalog # 30623-014
Supplier: Chemglass
Supplier #: CLS-1480-13SC
Jump to:
Product Details & Documents
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Specifications
- Color:Schott Blue
- Description:Solid Caps
- Thread size:GL45