Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Orange, GL32
Catalog # 30623-012
Supplier: Chemglass
Supplier #: CLS-1480-12
Jump to:
Product Details & Documents
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Specifications
- Color:Orange
- Description:Solid Caps
- Thread size:GL32