VWR® Heated Circulating Baths
Product Details & Documents
VWR® Heated Circulating Baths offer precise temperature control, at temperature ranges from ambient +10° to 200°C (depending on the controller).
- Available in a wide range of sizes
- Three controller options
- Ideal for many applications
The large, LCD display on the MX temperature controller is easily viewed across the lab
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
Certifications: ETL listed. CE marked. MX Controller complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. Advanced Digital and Advanced Programmable comply with DIN 12876-1 Class III safety requirements for use with flammable liquids. cETLus compliant.
Ordering information: Heated Circulators include reservoir cover; bypass tubing; male inlet and outlet adapters for 3/8, 1/4" and 3/16" (9.5, 6.3 and 4.7 mm ) tubing. 1/4" NPT to M16 adapters also included on 50 Hz models.
Services: Every VWR® ISO 17025 NIST Traceable Pre-calibrated Circulating Bath is calibrated and tested by an ISO 17025 accredited lab to ensure the highest standard of accuracy and traceability. Two options are available: Option 1: Standard Pre-calibrated circulating bath is calibrated and tested at both 37°C and 70°C. Option 2: Customer specified pre-calibrated circulating bath provides a choice of 3 different temperature points based on your research requirements. Simply order the stated catalog number and you will receive a protective box containing a work order form allowing you to specify your own calibration temperature points. After completing the work order, you can mail the box using a pre-paid return shipping label or you can scan and email the information to a specified E-mail address. The Avantor calibration team will calibrate the circulating bath based on your instruction.
VWRCATALYST™ can provide an array of services, such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWR[email protected] for further details.
Documents
89202-930
Images 360
App Notes Biotech Indus Pharm AppNotes E&MR
Product Literature
Circulating Bath
Videos
Specifications
- Temperature range:Ambient +10 to 135 °C
- Description:20 L Heated Circulating Bath, MX Controller
- Cat. No.:89400-978
- Max. pressure:2.3 psi (0.16 bar)
Complementary Products
Product Information
- Catalog #DescriptionProbeRangeAvailabilityPrice
- 89369-150Platinum high-accuracy freezer themometerStainless-steel probe with handle−100.00...+199.99 °C (−148.00...+391.99 °F)Log in for availabilityEach - $574.74
Accessories
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- 6" External Pt100 Probe with 3 m (10') length cable6" External Pt100 Probe with 3 m (10') length cable89201-004VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationLog in for availabilityEach - $743.10
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- 6" External Pt100 Probe with 7.6 m (25') length cable6" External Pt100 Probe with 7.6 m (25') length cable89201-006VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationLog in for availabilityEach - $1,029.25
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- 6" External Pt100 Probe with 15 m (50') length cable6" External Pt100 Probe with 15 m (50') length cable89201-008VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersFor Monitoring the Temperature of an External ApplicationLog in for availabilityEach - $1,552.33
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- RS232 CableRS232 Cable89201-010VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersCable Length: 9'Log in for availabilityEach - $101.37
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- Ethernet CableEthernet Cable89201-012VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersCable Length: 9'Log in for availabilityEach - $88.59
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- USB-A to Mini-B CableUSB-A to Mini-B Cable89201-016VWR Refrigerated Circulating Baths and VWR Heated Circulating Baths with Advanced Digital and Advanced Programmable ControllersCable Length: 9'Log in for availabilityEach - $63.39
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- 1/4" ID Buna N Tubing1/4" ID Buna N Tubing89201-056VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems–40...120 °C, Order in Continuous Lengths Of 3' (1 m)Log in for availabilityEach - $33.01
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- Insulation for 1/4" TubingInsulation for 1/4" Tubing89201-062VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsOrder in Continuous Lengths of 2 Yards (6 ft.)Log in for availabilityEach - $143.30
Product Information
- Catalog #For Use WithDetailsAvailabilityPrice
- Tube ClampsTube Clamps89201-076VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath SystemsFor 1/4 and 3/8" ID Tubing, Stainless SteelLog in for availabilityEach - $16.46