Order Entry
Canada
Orders LinkContactUsLinkComponent
 

VWR® Buffer Solutions

Poly bottle, 500 mL, 1.05, Colorless
Catalog # CA11020-926
Supplier:   VWR
 :  15200-320
VWR® Buffer Solutions
Catalog # CA11020-926
Supplier:  VWR
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions

Poly bottle, 500 mL, 1.05, Colorless

CAS Number:  
Price
$297.04
Log in for availability
Each (500ml) Case of 6 (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These buffer solutions are for calibrating pH meters

They are traceable to NIST standards.

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Color:
    Colorless
  • Size:
    500 mL
  • pH:
    1.05

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.