VWR® Buffer Solutions
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.
Documents
Certificates
Search by Lot Numberspec_71006-730 spec_71006-744 spec_71006-768 spec_71006-774 spec_71006-810
Product specification
Specifications
Specifications
- Pack type:Cubetainer®
- Color:Colorless
- Size:10 L
- pH buffer type:Phosphate
- pH:11.00
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



