Sample Cups, Polystyrene, Dynalon
4.0 ml, 39 mm
30620-272
: Dynalon
: 202134
Clear polystyrene cups for use with older auto analyzers.
Also useful for general micro use. Conical bottom. 26mm height. LDPE Stopper available
- Capacity:4.0 ml
- Height:39 mm