Order Entry
Canada
Orders LinkContactUsLinkComponent
Sample Cups, Polystyrene, Dynalon

Sample Cups, Polystyrene, Dynalon

2.0 ml, 26 mm
Catalog # 30620-270
Supplier:   Dynalon
Supplier #:  202094
Sample Cups, Polystyrene, Dynalon
Catalog # 30620-270
Supplier:  Dynalon
Sample Cups, Polystyrene, Dynalon
Sample Cups, Polystyrene, Dynalon

2.0 ml, 26 mm

Price
$253.18
Log in for availability
Case of 1,000
Product Details & Documents

Clear polystyrene cups for use with older auto analyzers.

Also useful for general micro use. Conical bottom. 26mm height. LDPE Stopper available

Specifications
  • Capacity:
    2.0 ml
  • Height:
    26 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.