Polypropylene Bags, Dynalon
1.5 mil., 102×254 mm (4×10")
Catalog # 30620-110
Supplier: Dynalon
Supplier #: 628515
Jump to:
Product Details & Documents
Transparent general-use plastic bags.
- 0.0015" thick
- Polypropylene construction
Economical general-use plastic bags are especially useful as specimen, sample, and freezing bags and are ideal for storing and shipping items.
Specifications
- Thickness:1.5 mil.
- W×L:102×254 mm (4×10")



