Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Cupel Fork, Webber's Welding

Cupel Fork for #6A Cupels, 8W x 6D
Catalog # CA11019-812
Supplier:   Webber's Welding
 :  FA116S
Cupel Fork, Webber's Welding
Catalog # CA11019-812
Supplier:  Webber's Welding
Cupel Fork, Webber's Welding

Cupel Fork for #6A Cupels, 8W x 6D

Price
$674.79
Log in for availability
Each
Product Details & Documents

Cupel Fork is specially designed for heavy-duty applications against heat from ovens or furnaces, while its mild steel prongs provide durable and dependable service.

Ordering information: Overall length: 57"
Handle length: 46"

Specifications
  • Format:
    8W x 6D
  • Description:
    Cupel Fork for #6A Cupels