Order Entry
Canada
Orders LinkContactUsLinkComponent
Crucible Loading Fork, Webber's Welding

Crucible Loading Fork, Webber's Welding

For use with 40 g Crucibles, Fits 5 crucibles
Catalog # CA11019-810
Supplier:   Webber's Welding
Supplier #:  029-803-11
Crucible Loading Fork, Webber's Welding
Catalog # CA11019-810
Supplier:  Webber's Welding
Crucible Loading Fork, Webber's Welding

For use with 40 g Crucibles, Fits 5 crucibles

Price
$627.97
Log in for availability
Each
Product Details & Documents

Crucible Fork is specially designed for heavy-duty applications with user safety in mind.

  • Aluminum pipe handle helps insulate against heat from ovens or furnaces
  • Mild steel crucible holders provide durable and dependable service

Ordering information: Overall Length: 50"
Handle Length: 46"

Specifications
  • Format:
    Fits 5 crucibles
  • Description:
    For use with 40 g Crucibles