Order Entry
Canada
Orders LinkContactUsLinkComponent
Molybdate 3 Reagent Solution, Hach

Molybdate 3 Reagent Solution, Hach

Molybdate 3 Reagent Solution, 50 mL Self-Contained Dropping Bottle, Heteropoly blue, 50
Catalog # CA11016-694
Supplier:   Hach
Supplier #:  199526
Molybdate 3 Reagent Solution, Hach
Catalog # CA11016-694
Supplier:  Hach
CAS Number:  
Molybdate 3 Reagent Solution, Hach
Molybdate 3 Reagent Solution, Hach

Molybdate 3 Reagent Solution, 50 mL Self-Contained Dropping Bottle, Heteropoly blue, 50

CAS Number:  
Price
$92.65
Log in for availability
Each (50ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For low and ultra-low range silica determination by the Heteropoly Blue method

Hach methods 8282 and 8186.

Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.

Specifications

Specifications

  • Chemical method:
    Heteropoly blue
  • Description:
    Molybdate 3 Reagent Solution
  • Size:
    50 mL Self-Contained Dropping Bottle
  • No. of tests:
    50
  • Cat. No.:
    CA11016-694
  • Method:
    Heteropoly Blue

Specifications