Order Entry
Canada
Orders LinkContactUsLinkComponent
Variable Volume Pipettes, Hach

Variable Volume Pipettes, Hach

Variable Volume Pipet, 0.2 - 1.0 ml
Catalog # CA11014-356
Supplier:   Hach
Supplier #:  BBP078
Variable Volume Pipettes, Hach
Catalog # CA11014-356
Supplier:  Hach
Variable Volume Pipettes, Hach
Variable Volume Pipettes, Hach

Variable Volume Pipet, 0.2 - 1.0 ml

Price
$1,346.66
Log in for availability
Each
Product Details & Documents

Hach's variable volume single-channel, air-displacement pipettes feature robust design, comfortable ergonomics, slim-line tip ejection, and single-handed operation.

Outstanding accuracy with adjustments in 50 L increments (0.05 ml).

Ordering information: Supplied with calibration report and maintenance tool.

Specifications
  • Volume:
    0.2 - 1.0 ml
  • Description:
    Variable Volume Pipet
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.