Corning® Polypropylene Erlenmeyer Flasks, Corning
500 ml, 100 ml, GL-52
Supplier: Corning
Supplier #: 4985P-500
Jump to:
Product Details & Documents
PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.
Documents
Specifications
- Graduation Interval:100 ml
- Capacity:500 ml
- Thread:GL-52
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



