Laemmli SDS sample buffer, non-reducing (4X), Thermo Scientific
50 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J63615-AD
Laemmli SDS sample buffer, non-reducing (4X), Thermo Scientific
Catalog # CAAAJ63615-AD
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system
Documents
Specifications
Specifications
- Size:50 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



