Order Entry
Canada
Orders LinkContactUsLinkComponent
Research Buffers and Reagents, RNase and DNase free, G-Biosciences

Research Buffers and Reagents, RNase and DNase free, G-Biosciences

CTAB Extraction Buffer, 60 ml
Catalog # CA89167-780
Supplier:   G-Biosciences
Supplier #:  786-564
Research Buffers and Reagents, RNase and DNase free, G-Biosciences
Catalog # CA89167-780
Supplier:  G-Biosciences
CAS Number:  
Research Buffers and Reagents, RNase and DNase free, G-Biosciences

CTAB Extraction Buffer, 60 ml

CAS Number:  
Price
$95.41
Log in for availability
Each (60ml)
Product Details & Documents

G-Biosciences offers a selection of DNase and RNase free buffers designed to ensure high quality research.

Ordering information: Accessories available.

Specifications

Specifications

  • Description:
    CTAB Extraction Buffer
  • Size:
    60 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.