Orders LinkContactUsLinkComponent
Dextran sulfate
Bottle, 250 g
  CA71003-394
 :  RC-044
Dextran sulfate
  CA71003-394
 :  9011-18-1
Dextran sulfate
Bottle, 250 g
 :  9011-18-1
$3,288.20
Each (250g)
Dextran sulfate sodium (CAS 9011‑18‑1) is a versatile high‑molecular‑weight polyanion, essential in molecular biology (e.g., hybridization), biochemistry (e.g., protein/lipoprotein precipitation), virology, inflammation modeling, and nanomedicine construction.
  • White to off-white crystalline powder BSE/TSE Free Loss on Drying: ≤10 % Free Sulfate: ≤0.5 % pH (1%, Water) at 25 °C: 5.0 to 7.5 Chloride: ≤1000 ppm Sulfur Content: 17.0 to 20.0 %
 :  Dextran sulfate sodium from Dextran, Dextran sulphate sodium from Dextran
  •  :9011-18-1
  •  :>500.000 g/mol
  • Pack type:
    Bottle
  • Size:
    250 g
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.