Orders LinkContactUsLinkComponent
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
High Carbon Ferro-Chrome Single Std
  102099-012
 :  80074-BAS-FC
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
  102099-012
 :  
 :  
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
High Carbon Ferro-Chrome Single Std
 :  
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. Supplied in 2, 3, and 5 mm diameter and a location map for standard identification. They are supplied with fully authenticated certificates of analysis.
  • Description:
    High Carbon Ferro-Chrome Single Std
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.