Orders LinkContactUsLinkComponent
Micro Scissors, Rubis, Electron Microscopy Sciences
Style Micro Scissors, 1C200, 146 mm (5¾"), 6.4 mm (¼")
  102098-976
 :  78180-1C2
Micro Scissors, Rubis, Electron Microscopy Sciences
  102098-976
Micro Scissors, Rubis, Electron Microscopy Sciences
Micro Scissors, Rubis, Electron Microscopy Sciences
Style Micro Scissors, 1C200, 146 mm (5¾"), 6.4 mm (¼")
81,31 $
Each
Micro scissors manufactured from hardened, anti-acid, stainless, surgical steel (CA)
Rubis Micro Scissors Set includes one light duty Rubis Colibri fine scissors, one medium duty Rubis Classic medium fine scissors, and one heavy duty Rubis Sauro medium strong scissors. Strong, slightly curved easily cut through small bones and cartilages.
Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist® grip scissors offer ergonomically, comfort and control.
  • Description:
    Style Micro Scissors, 1C200
  • Length:
    146 mm (5¾")
  • Blade length:
    6.4 mm (¼")
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)