Orders LinkContactUsLinkComponent
Labeling Tape, Electron Microscopy Sciences
Green, 0.75" x 500"
  102098-758
 :  77627-G
Labeling Tape, Electron Microscopy Sciences
  102098-758
Labeling Tape, Electron Microscopy Sciences
Labeling Tape, Electron Microscopy Sciences
Green, 0.75" x 500"
38,31 $
Each Roll of 1
Use pen, pencil or marker to mark. Leaves no adhesive residue when peeled off.
Water, oil, and acid resistant. Withstands cold (−23 °C) and hot (121 °C) temperatures.
  • Color:
    Green
  • Size:
    0.75" x 500"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)