Specialty Labels, Electron Microscopy Sciences
Product Details & Documents
Preprinted Tough-Spots make labeling and identifying samples quick and easy.
- Temperature resistant from –196 to 80 °C
Use the numbered Tough-Spots to identify a series of samples - no handwriting or printing needed. Each color coded ToughSpot has a distinctly different color to identify similar samples and also includes a white area to clearly write a quick note about it.
Lab safety is always a top priority. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.
Specifications
- Color:Copper
- Description:Tough-Spots
- Legend:Bullseye
- Diameter:7/16"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.