Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Product Details & Documents
Tough-Spots® Labels are pre-cut peel-off labels specially for microcentrifuge tube tops. This chemically resistant, flexible labels that can withstand boiling, freezing to −196 °C, sustained humidity.
- Polyolefin labels resist most organic solvents and caustic agents
- They adhere to most plastics, glass and metals without cracking, peeling, or degrading
- Heat-resistant, stay-flat sheets will not jam in laser printers
- Label sheets are top-coated to readily accept laser printing
- Assortment includes four sheets of blue, green, red, yellow and orange color
Made of polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
Specifications
- Color:Blue
- Description:Adhesive laser labels, Tough-Spots, round
- Packaging:3840 labels/pack
- Diameter:1/2"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)