Adhesive Labels on Rolls, Round, Electron Microscopy Sciences
Product Details & Documents
Tough-Spots® pre-cut peel-off round labels are used on microcentrifuge tube tops. The PVC labels are easy to write on and accept writing from most lab markers including solvent resistant pens and teeny writers.
- In sheets (for laser printer) or on a roll
- Select from 11 colors
- A variety of sizes, colors and formats ensures the right label for the job
Ordering information: Rainbow includes blue, green, orange, red, and yellow.
Delivery information: 5 Across Tough-Spots® come in a dispensing box that assists in labeling individual tubes used in small experiments. White or assorted roll with 1000 spots each of blue, green, lavender, red and yellow.
Specifications
- Color:Lavender
- Description:Labels on rolls
- Dimensions:0.5"
- Packaging:1000 labels/roll
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.