Orders LinkContactUsLinkComponent
Variable Tilt Mount holder, Electron Microscopy Sciences
Variable Tilt Mount holder, Electron Microscopy Sciences
Variable Tilt Mount, M4 Hitachi
Catalog # 102098-302
Supplier #:  75952-05H
Variable Tilt Mount holder, Electron Microscopy Sciences
Catalog # 102098-302
Supplier:  
Variable Tilt Mount holder, Electron Microscopy Sciences
Variable Tilt Mount holder, Electron Microscopy Sciences
Variable Tilt Mount, M4 Hitachi
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This mount which is made from aluminum is used for the mounting and tilting of samples from 0° – 90° as well as for SEM investigation with small working distances. The mount is marked for 30°, 45°, 70° and 90°. The table measures 11 x 11 mm (0.46” x0.46”)and the overall size is 14 x 14 x 12.7 mm.
Specifications
  • Description:
    Variable Tilt Mount, M4 Hitachi
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)