Orders LinkContactUsLinkComponent
Mini SEM Mount, Electron Microscopy Sciences
Mini Pin, Slotted Head, 6.6 x 1.3 mm
  102098-236
 :  75638-30
Mini SEM Mount, Electron Microscopy Sciences
  102098-236
Mini SEM Mount, Electron Microscopy Sciences
Mini SEM Mount, Electron Microscopy Sciences
Mini Pin, Slotted Head, 6.6 x 1.3 mm
$46.20
Each Pack of 100
Mini Pin, Aluminum For FEI, Tescan, Zeiss, Philips, Leo, Cambridge, AMRAY, Leica, CamScan, ETEC.
Head: 6.6 x 1.3 mm, Pin: 9.5 mm.
  • Description:
    Mini Pin, Slotted Head
  • Size:
    6.6 x 1.3 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.