Orders LinkContactUsLinkComponent
Mini SEM Mount, Electron Microscopy Sciences
Mini Pin, Slotted Head, 6.6 x 1.3 mm
Catalog # 102098-234
 :  75638-20
Mini SEM Mount, Electron Microscopy Sciences
Catalog # 102098-234
Supplier:  
Mini SEM Mount, Electron Microscopy Sciences
Mini SEM Mount, Electron Microscopy Sciences
Mini Pin, Slotted Head, 6.6 x 1.3 mm
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mini Pin, Aluminum For FEI, Tescan, Zeiss, Philips, Leo, Cambridge, AMRAY, Leica, CamScan, ETEC.
Head: 6.6 x 1.3 mm, Pin: 9.5 mm.
Specifications
  • Description:
    Mini Pin, Slotted Head
  • Size:
    6.6 x 1.3 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)