Histo-Mark Pen, Electron Microscopy Sciences
Product Details & Documents
The fine-line Histo-Mark pen is designed to work like a pen.
- Can be used on any surface
- Marking quality remains consistent throughout the life of the ink barrel
The Histo-Mark pen is solvent resistent, especially to alcohol, acetone, xylene, xylene subsite and toluene.
Specifications
- Point type:Fine
- Color:Black
- Description:Histo-Mark pen, set of 12
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.