Orders LinkContactUsLinkComponent
Scalpel Blades, Electron Microscopy Sciences
Premiere™ Blade #15
Catalog # 102097-834
 :  72049-15
Scalpel Blades, Electron Microscopy Sciences
Catalog # 102097-834
Scalpel Blades, Electron Microscopy Sciences
Scalpel Blades, Electron Microscopy Sciences
Premiere™ Blade #15
Price
55,29 $
Log in for availability
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Premiere® Blades are made from carbon steel, individually foil wrapped and gamma radiation sterilized.
They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Specifications
  • Description:
    Premiere™ Blade #15
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.