Orders LinkContactUsLinkComponent
Pop-Up Slide Holder, Electron Microscopy Sciences
Pop-Up Slide Holder, Electron Microscopy Sciences
30 Slide Pop-Up Slide Folder
Catalog # 102097-588
Supplier #:  71522-10
Pop-Up Slide Holder, Electron Microscopy Sciences
Catalog # 102097-588
Pop-Up Slide Holder, Electron Microscopy Sciences
Pop-Up Slide Holder, Electron Microscopy Sciences
30 Slide Pop-Up Slide Folder
Price
$364.65
Log in for availability
Case of 10 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Pop-Up holder holds 20 or 30 of 1 x 3" microscope slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed. Measurements: 8W x 131/2L x 11/32H".
Specifications
  • Description:
    30 Slide Pop-Up Slide Folder
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.