Orders LinkContactUsLinkComponent
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Dial A Grid-N-Block storage box
Catalog # 102097-426
Supplier #:  71158-10
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Catalog # 102097-426
Supplier:  
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
EMS Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Dial A Grid-N-Block storage box
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Modules have 24 slots labeled with letters from A-X where the grids can be stored and rotated for easy access as well as 3 additional cavities for block storage .
Available with and without a unique identification number.
Specifications
  • Description:
    Dial A Grid-N-Block storage box
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)