Orders LinkContactUsLinkComponent
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Replacement Blades for Dispenser
Catalog # 102097-224
Supplier #:  70993-04
Parafilm Dispenser, Electron Microscopy Sciences
Catalog # 102097-224
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Replacement Blades for Dispenser
Price
$27.14
Log in for availability
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags. Unique design for maximum safety. ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to your fingers.
  • Holds one or two 2" rolls, or one 4" roll
  • Size (W x H x D): 120 x 156 x 171 mm
  • Parafilm is a registered trademark of American can company
Specifications
  • Description:
    Replacement Blades for Dispenser
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)