Orders LinkContactUsLinkComponent
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Two counting unit tally counter
Catalog # 102096-604
Supplier #:  70310-02
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Catalog # 102096-604
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Two counting unit tally counter
Price
$482.66
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The desk tally counter comes with one, two, or six counting units.
One Counting Unit: A chrome plated with counter with a flange (21/2" dia.) for desktop mounting. The recording lever is on the right side and the reset knob on the top. The counter is ideal for serial sectioning work. Four digit place.
Two Counting Units: A Counter with two counting keys each with 0 to 999 range with a single reset knob it comes with removable tabs for top identification.
Six Counting Units: A counter with five keys and one totalize unit (0 to 999), which records the total number of strokes made on other keys. At 100 increments, totalizing unit rings bell. When totalizing unit records 100, each figure corresponding to individual keys automatically displays percentage. Two reset knobs, removable tabs on top of counter provide area for identification of clasification being counted.
Specifications
  • Description:
    Two counting unit tally counter
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.