Orders LinkContactUsLinkComponent
Cutting Mats, Electron Microscopy Sciences
Cutting Mats, Electron Microscopy Sciences
Cutting Mat 2.5 x 3"
Catalog # 102096-430
Supplier #:  69037-23
Cutting Mats, Electron Microscopy Sciences
Catalog # 102096-430
Cutting Mats, Electron Microscopy Sciences
Cutting Mats, Electron Microscopy Sciences
Cutting Mat 2.5 x 3"
Price
$25.95
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These cutting mats are inert, self-healing and can be sterilized with ethanol. Available sizes 6 x 8" and 21/2 x 3".
Specifications
  • Description:
    Cutting Mat 2.5 x 3"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)