Gel Drying Frames, Electron Microscopy Sciences
Product Details & Documents
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers.
- Two open frames
- Two solid back plates
- 12 side clips
- 50 sheets of pre-cut cellophane
The 24 x 24 cm frames allow for drying of larger format gels, while the 14 x 14 cm frames are ideal for mini gels and other smaller formats.
Specifications
- Description:Gel Drying Frames 24 x 24 cm Gel Frame
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.