Orders LinkContactUsLinkComponent
Dounce Tissue Grinder, Electron Microscopy Sciences
Dounce Tissue Grinder, 1 ml
Catalog # 102095-538
 :  64790-01
Dounce Tissue Grinder, Electron Microscopy Sciences
Catalog # 102095-538
Dounce Tissue Grinder, Electron Microscopy Sciences
Dounce Tissue Grinder, Electron Microscopy Sciences
Dounce Tissue Grinder, 1 ml
Price
$956.24
Log in for availability
Each Pack of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This grinder designed to retain a high percentage of cell nuclei and mitochondria in soft tissues from cell cultures. When use, the pestle ball is encircled in liquid, which is avoid heat buildup by reducing friction.
  • Ideal for soft tissue
  • Provide with loose and tight pestle
  • Wheaton 33 low extractable borosilicate glass
  • Autoclavable
This grinder is ideal for enzyme studies. The initial grinding is performed using the “loose” pestle. The grinding process is completed using the “tight” pestle.. This grinder is used by moving the pestle up and down, and work best with cell suspensions or very soft tissues. The mortar has a large reservoir and pouring lip and is supplied with “loose” and “tight” pestles. Tight Pestle “A” Specification: 0.0010 – 0.0030” (0.025 – 0.076 mm) Loose Pestle “B” Specification: 0.0035 – 0.0055” (0.089 – 0.14 mm)
Specifications
  • Description:
    Dounce Tissue Grinder
  • Size:
    1 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.