Orders LinkContactUsLinkComponent
MeshWell Plates, Electron Microscopy Sciences
MeshWell Plates, Electron Microscopy Sciences
Plate 12–well with Cluster
  102095-516
Supplier #:  64702-12
MeshWell Plates, Electron Microscopy Sciences
  102095-516
MeshWell Plates, Electron Microscopy Sciences
MeshWell Plates, Electron Microscopy Sciences
Plate 12–well with Cluster
$574.34
Each
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500 µm opening.
  • Available with 6, 12, and 24 well configurations
  • Also able to process tissue slices with Reagent Trays
Fits into the 6, 12 and 24 Costar/Corning Cell Culture Clusters for processing tissue slices.
Ordering information: The MeshWell Plate set includes one MeshWell and one Cluster plate.
  • Description:
    Plate 12–well with Cluster
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.