MeshWell Plates, Electron Microscopy Sciences
Product Details & Documents
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500 µm opening.
- Available with 6, 12, and 24 well configurations
- Also able to process tissue slices with Reagent Trays
Fits into the 6, 12 and 24 Costar/Corning Cell Culture Clusters for processing tissue slices.
Ordering information: The MeshWell Plate set includes one MeshWell and one Cluster plate.
Specifications
- Description:Plate 6–well with Cluster
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)