Tri-Seal™ container, Electron Microscopy Sciences
Product Details & Documents
Tri-Seal™ Specimen Container is a patented, leakproof, airtight, touch-top specimen container for the pathology lab.
Vials have permanent hinge and triple seal design. These are molded from 100% High-Density, translucent polypropylene and are available in a variety of sizes from 13 ml to 300 ml.
Specifications
- Capacity:300 ml
- Description:Tall With Locking Latch
- Dimensions:30 × 80 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.